Merck
  • Cross-linking of C-terminal residues of phospholamban to the Ca2+ pump of cardiac sarcoplasmic reticulum to probe spatial and functional interactions within the transmembrane domain.
Cross-linking of C-terminal residues of phospholamban to the Ca2+ pump of cardiac sarcoplasmic reticulum to probe spatial and functional interactions within the transmembrane domain.
The Journal of biological chemistry (2006-03-24)
Zhenhui Chen, Brandy L Akin, David L Stokes, Larry R Jones
RESUMEN
Interactions between the transmembrane domains of phospholamban (PLB) and the cardiac Ca2+ pump (SERCA2a) have been investigated by chemical cross-linking. Specifically, C-terminal, transmembrane residues 45-52 of PLB were individually mutated to Cys, then cross-linked to V89C in the M2 helix of SERCA2a with the thiol-specific cross-linking reagents Cu2+-phenanthroline, dibromobimane, and bismaleimidohexane. V49C-, M50C-, and L52C-PLB all cross-linked strongly to V89C-SERCA2a, coupling to 70-100% of SERCA2a molecules. Residues 45-48 and 51 of PLB also cross-linked to V89C of SERCA2a, but more weakly. Evidence for the mechanism of PLB regulation of SERCA2a was provided by the conformational dependence of cross-linking. In particular, the required absence of Ca2+ for cross-linking implicated the E2 conformation of SERCA2a, and its enhancement by ATP confirmed E2 x ATP as the conformation with the highest affinity for PLB. In contrast, E2 phosphorylated with inorganic phosphate (E2P) and E2 inhibited by thapsigargin (E2 x TG) both failed to cross-link to PLB. These results with transmembrane PLB residues are completely consistent with cytoplasmic PLB residues studied previously, suggesting that the dissociation of PLB from the Ca2+ pump is complete, not partial, when the pump binds Ca2+ (E1 x Ca2) or adopts the E2P or E2 x TG conformations. V49C of PLB cross-linked to 100% of SERCA2a molecules, suggesting that this residue might have functional importance for regulation. Indeed, we found that mutation of Val49 to smaller side-chained residues V49A or V49G augmented PLB inhibition, whereas mutation to the larger hydrophobic residue, V49L, prevented PLB inhibition. A model for the interaction of PLB with SERCA2a is presented, showing that Val49 fits into a constriction at the lumenal end of the M2 helix of SERCA, possibly controlling access of PLB to its binding site on SERCA.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dibromobimane, BioReagent, suitable for fluorescence, ≥95.0% (CHN)