Merck
Functional subtypes of renal alpha1-adrenoceptor in diabetic and non-diabetic 2K1C Goldblatt renovascular hypertension.
Acta pharmacologica Sinica (2008-04-24)
A Armenia, Munavvar Abdul Sattar, Nor Azizan Abdullah, Md Abdul Hye Khan, Edward James Johns
RESUMEN
This study investigates the subtypes of the alpha1-adrenoceptor mediating the adrenergically-induced renal vasoconstrictor responses in streptozotocin-induced diabetic and non-diabetic 2-kidney one clip (2K1C) Goldblatt hypertensive rats. The renal blood flow responses to renal nerve stimulation, noradrenaline, phenylephrine, and methoxamine were measured in the absence and presence of nitrendipine, 5-methylurapidil, chloroethylclonidine and BMY 7378. The renal vasoconstrictor responses were markedly attenuated by nitrendipine and 5- methylurapidil in the diabetic rats (all P< 0.05). In the non-diabetic rats, these responses were markedly attenuated by nitrendipine, 5-methylurapidil, and BMY 7378 (all P< 0.05). In both experimental groups, chloroethylclonidine markedly accentuated the renal vasoconstrictions caused by all the adrenergic stimuli (all P< 0.05). These observations indicate that alpha 1A-adrenoceptor subtypes play a major role in mediating adrenergically-induced renal vasoconstriction in the diabetic 2K1C Goldblatt hypertensive rats. In the non-diabetic 2K1C Goldblatt hypertensive rats, contributions of alpha 1A and alpha 1D-adrenoceptor subtypes were proposed. Apart from post-synaptic alpha 1-adrenoceptors, both in the diabetic and non-diabetic 2K1C Goldblatt hypertensive rats, the potential involvement of presynaptic alpha 1- adrenoceptors is also suggested.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
5-Methylurapidil, solid