Merck
[Stability of the tetramine in blood samples under different temperatures].
Fa yi xue za zhi (2009-12-17)
Jun Bu, Wei Liu, Hui Yan
RESUMEN
To study the stability of tetramine in blood samples under different environment temperatures. The tetramine samples in 0.5 microg/mL concentration were stored in the environment temperature at 45 degrees C, 25 degrees C and 4 degrees C. The tetramine concentration were measured by GC-MS in 1, 3, 12, 18 and 39 days, respectively. And then the results of measurement were analyzed by SPSS software. At 45 degrees C and 25 degrees C, the concentration of tetramine changed without distinctly in first 3 days and declined significantly from 4 days to 12 days. Thereafter it declined more slowly. At 4 degrees C, the concentration of tetramine changed without distinctly in 12 days and declined significantly from 13 days to 18 days. Thereafter it declined more slowly. The concentration of tetra mine in blood samples is stable in 3 days. Thereafter it declined gradually. Temperature also influences the concentration of tetra mine.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Tetramethylenedisulfotetramine solution, 100 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®