Merck
Excited state proton transfer in the Cinchona alkaloid cupreidine.
Physical chemistry chemical physics : PCCP (2010-08-21)
Junhong Qian, Albert M Brouwer
RESUMEN
Photophysical properties of the organocatalyst cupreidine (CPD) and its chromophoric building block 6-hydroxyquinoline (6HQ) in protic and nonprotic polar solvents (methanol and acetonitrile) were investigated by means of UV-vis absorption, and steady state and time resolved fluorescence spectroscopy. The effects of the catalytically relevant interactions with electrophilic and hydrogen bonding agents (p-toluene sulfonic acid and water) on their spectral characteristics were studied. In neutral CPD in acetonitrile, quenching of fluorescence occurs due to electron transfer from the quinuclidine nitrogen to the excited quinoline chromophore. Protonation suppresses this process, while complexation with water leads to enhanced excited state proton transfer from the 6'-OH group to the quinuclidine nitrogen, and emission occurs from the anionic form of the chromophore. The weakly emitting zwitterionic form of the hydroxyquinoline chromophore is readily formed in methanol, but not in acetonitrile.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
6-Hydroxyquinoline, 95%