Merck
  • Active cyamemazine metabolites in patients treated with cyamemazine (Tercian®): influence on cerebral dopamine D2 and serotonin 5-HT (2A) receptor occupancy as measured by positron emission tomography (PET).
Active cyamemazine metabolites in patients treated with cyamemazine (Tercian®): influence on cerebral dopamine D2 and serotonin 5-HT (2A) receptor occupancy as measured by positron emission tomography (PET).
Psychopharmacology (2011-04-12)
Yann Hodé, Amine Benyamina, Christophe Arbus, Matthias Reimold
RESUMEN
Cyamemazine (Tercian®) is an antipsychotic agent blocking central dopamine D(2) receptors, which induces few extrapyramidal adverse effects, due to a potent antagonistic action at serotonin 5-HT(2A) receptors. In vitro studies showed that the desmethyl metabolite of cyamemazine (N-desmethyl cyamemazine) has similar affinity for 5-HT(2A) receptors as cyamemazine, whereas its D(2) receptor affinity is eight times lower (Benyamina et al. in Eur J Pharmacol 578(2-3):142-147, 2008). Moreover, cyamemazine sulfoxide showed modest affinity for 5-HT(2A) receptors. The objective of this study is to measure steady-state plasma levels of N-desmethyl cyamemazine and cyamemazine sulfoxide in patients treated with clinically relevant doses of cyamemazine and correlate them with dopamine D(2) and serotonin 5-HT(2A) receptor occupancies (RO) assessed by positron emission tomography (PET). Eight patients received Tercian® 37.5, 75, 150, or 300 mg/day according to their symptoms. Dopamine D(2) and serotonin 5-HT(2A) RO were assessed at steady-state cyamemazine plasma levels using [(11)C]raclopride and [(11)C]N-methyl-spiperone, respectively, for PET. Plasma levels of cyamemazine metabolites were determined using a validated high-performance liquid chromatography (PerkinElmer) associated with a mass spectrometry detection (API 365, PE SCIEX). The apparent equilibrium inhibition constant (K (i)) was estimated by fitting RO with plasma levels of cyamemazine metabolites at the time of the PET scan. After 6 days of cyamemazine administration, plasma N-desmethyl cyamemazine reached steady-state levels at 2 to 12 times higher than those previously found for cyamemazine (Hode et al. in Psychopharmacology (Berl) 180:377-384, 2005). Plasma levels of N-desmethyl cyamemazine were closely related to striatal D(2) RO (r (2) = 0.942) and extrastriatal 5-HT(2A) RO (r (2) = 0.901). The estimated K (i(app)) value of N-desmethyl cyamemazine for striatal D(2) receptors was about fivefold higher than that for extrastriatal 5-HT(2A) receptors (48.7 vs. 10.6 nM). Striatal D(2) RO increased with the plasma levels of N-desmethyl cyamemazine but remained below 75% even at its highest levels. At steady state, plasma cyamemazine sulfoxide levels were about double those of N-desmethyl cyamemazine. However, these cyamemazine sulfoxide levels should not contribute significantly to in vivo 5-HT(2A) and D(2) receptor occupancy. In patients orally given cyamemazine, N-desmethyl cyamemazine, but not cyamemazine sulfoxide, should significantly contribute to in vivo frontal 5-HT(2A) and striatal D(2) receptor occupancy. The higher in vivo affinity of cyamemazine and its desmethyl metabolite for serotonin 5-HT(2A) receptors compared with dopamine D(2) receptors should explain the low incidence of extrapyramidal adverse effects.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cyamemazine, ≥98% (HPLC)