Merck
  • Helicobacter pylori cholesteryl glucosides interfere with host membrane phase and affect type IV secretion system function during infection in AGS cells.
Helicobacter pylori cholesteryl glucosides interfere with host membrane phase and affect type IV secretion system function during infection in AGS cells.
Molecular microbiology (2011-11-08)
Hung-Jung Wang, Wen-Chi Cheng, Hsin-Hung Cheng, Chih-Ho Lai, Wen-Ching Wang
RESUMEN
Helicobacter pylori infection is an aetiological cause of gastric disorders worldwide. H. pylori has been shown to assimilate and convert host cholesterol into cholesteryl glucosides (CGs) by cholesterol-α-glucosyltransferase encoded by capJ. Here, we show that CapJ-deficient (ΔcapJ) H. pylori resulted in greatly reduced type IV secretion system (TFSS)-associated activities, including the hummingbird phenotype of AGS cells, IL-8 production, CagA translocation/phosphorylation and CagA-mediated signalling events. Complementation of the ΔcapJ mutation with wild type cagJ or by adding CGs-containing lysates or exogenous fluorophore-tagged CGs reversed the mutant phenotypes. We also show that the wild-type but not ΔcapJ H. pylori recruited raft-associated components to sites of bacterial attachment. Fluorescence recovery after photobleaching (FRAP) analysis of AGS cells treated with fluorescence-tagged cholesterol/CGs revealed that there was a higher proportion of CGs associated with immobile fractions. CGs-associated membranes were also more resistant to a cold detergent extraction. Thus, we propose that CGs synthesized by H. pylori around host-pathogen contact sites partition in detergent-resistant membranes (DRMs), alters lateral-phase segregation in membrane and reorganizes membrane architecture. These processes together promote the formation of a functional TFSS and H. pylori infection.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cholesterol β-D-glucoside, ≥97% (TLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad