Merck
Si-free enolate Claisen rearrangements of enamido substrates.
Organic & biomolecular chemistry (2012-01-05)
Wesley R R Harker, Emma L Carswell, David R Carbery
RESUMEN
α-Alkyl β-amino esters are available in high diastereoselectivity through a silicon-free Claisen enolate [3,3]-sigmatropic rearrangement of enamide esters. Optimisation studies have probed the crucial role of the initial enolisation and the nature of the enamide N-centre. The demonstration of chirality transfer and the formation of β-proline systems, is also presented.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(S)-(+)-Pyrrolidine-3-carboxylic acid, ≥98.0% (NT)