Merck
  • Efficacy and safety of azelastine 0.15% nasal spray administered once daily in patients with allergy to Texas mountain cedar pollen.
Efficacy and safety of azelastine 0.15% nasal spray administered once daily in patients with allergy to Texas mountain cedar pollen.
International forum of allergy & rhinology (2012-01-31)
William C Howland, Niran J Amar, William Wheeler, Harry Sacks
RESUMEN
A previous study with azelastine nasal spray in patients with seasonal allergic rhinitis (SAR) demonstrated that increasing the azelastine concentration from 0.1% to 0.15% allowed for once-daily dosing without increasing the incidence of adverse effects. This study evaluated the efficacy of azelastine 0.15% nasal spray administered once daily for treating symptoms of SAR. In this 14-day, randomized, double-blind, placebo-controlled study, patients with moderate-to-severe SAR were randomized to azelastine 0.15% (n = 251) or placebo (n = 255), both at a dosage of 2 sprays/nostril once daily. The primary efficacy variable was change from baseline in the 12-hour reflective Total Nasal Symptom Score (TNSS). Key secondary variables were change from baseline in 24-hour instantaneous TNSS, to establish the dosing interval, and change from baseline in the Total Ocular Symptom Score (TOSS). The mean improvement (3.57) and percentage improvement (19.3%) in 12-hour reflective TNSS was significant (p < 0.012) with azelastine 0.15% compared to placebo (2.14 and 11.4%, respectively). The mean improvement in 24-hour instantaneous TNSS was also significant (p < 0.001) for azelastine 0.15% compared to placebo, indicating efficacy with once-daily dosing. The overall improvement and percentage improvement in TOSS was significant (p ≤ 0.012) with azelastine 0.15% (2.21 and 16.7%, respectively) compared to placebo (1.28 and 6.0%, respectively). The overall score for the Rhinoconjunctivitis Quality of Life Questionnaire (RQLQ) was significantly (p < 0.001) improved from baseline in the azelastine group compared with the placebo group. Nasal discomfort (3.6%) and bitter taste (2.4%) were the most common adverse events. There were no reports of somnolence with azelastine. Azelastine 0.15% was effective and well tolerated with once-daily dosing. Azelastine 0.15% nasal spray significantly improved a complex of eye symptoms compared to placebo.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Azelastine hydrochloride, ≥98% (HPLC)
Azelastine hydrochloride, European Pharmacopoeia (EP) Reference Standard