Merck
Long-term use of HU210 adversely affects spermatogenesis in rats by modulating the endocannabinoid system.
International journal of andrology (2012-03-23)
S E M Lewis, R Paro, L Borriello, L Simon, L Robinson, Z Dincer, G Riedel, N Battista, M Maccarrone
RESUMEN
Recent societal acceptance of cannabinoids as recreational and therapeutic drugs has posed a potential hazard to male reproductive health. Mammals have a highly sophisticated endogenous cannabinoid (ECS) system that regulates male (and female) reproduction and exo-cannabinoids may influence it adversely. Therefore it is imperative to determine their effects on male reproduction so that men can make informed choices as to their use. Here, an animal model was used to administer HU210, a synthetic analogue of Δ9-tetrahydrocannabinol (THC) and potent cannabinoid receptor (CB) agonist to determine its effects on reproductive organ weights, spermatogenesis, testicular histology and sperm motility. Its effects on the physiological endocannabinoid system were also investigated. Spermatogenesis was markedly impaired with reductions in total sperm count after 2 weeks of exposure. Spermatogenic efficiency was depleted, and Sertoli cell number decreased as exposure time increased with seminiferous tubules showing germ cell depletion developing into atrophy in some cases. Sperm motility was also adversely affected with marked reductions from 2 weeks on. HU210 also acted on the sperm's endocannabinoid system. Long-term use of exo-cannabinoids has adverse effects on both spermatogenesis and sperm function. These findings highlight the urgent need for studies evaluating the fertility potential of male recreational drug users. HU210, a selective agonist for CB1 and CB2 cannabinoid receptors impairs spermatogenesis and sperm motility and deregulates the endocannabinoid system.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
HU-210 solution, 100 μg/mL in methanol, ampule of 1 mL, (Spice Cannabinoid), certified reference material, Cerilliant®
Sigma-Aldrich
HU-210, solid (air sensitive)