Merck
Atl1 regulates choice between global genome and transcription-coupled repair of O(6)-alkylguanines.
Molecular cell (2012-06-05)
Vitaly F Latypov, Julie L Tubbs, Amanda J Watson, Andrew S Marriott, Gail McGown, Mary Thorncroft, Oliver J Wilkinson, Pattama Senthong, Amna Butt, Andrew S Arvai, Christopher L Millington, Andrew C Povey, David M Williams, Mauro F Santibanez-Koref, John A Tainer, Geoffrey P Margison
RESUMEN
Nucleotide excision repair (NER) has long been known to remove DNA lesions induced by chemical carcinogens, and the molecular mechanism has been partially elucidated. Here we demonstrate that in Schizosaccharomyces pombe a DNA recognition protein, alkyltransferase-like 1 (Atl1), can play a pivotal role in selecting a specific NER pathway, depending on the nature of the DNA modification. The relative ease of dissociation of Atl1 from DNA containing small O(6)-alkylguanines allows accurate completion of global genome repair (GGR), whereas strong Atl1 binding to bulky O(6)-alkylguanines blocks GGR, stalls the transcription machinery, and diverts the damage to transcription-coupled repair. Our findings redraw the initial stages of the NER process in those organisms that express an alkyltransferase-like gene and raise the question of whether or not O(6)-alkylguanine lesions that are poor substrates for the alkyltransferase proteins in higher eukaryotes might, by analogy, signal such lesions for repair by NER.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
6-O-Methylguanine, 97%