Merck
  • Tetrahydroindazolone substituted 2-aminobenzamides as fluorescent probes: switching metal ion selectivity from zinc to cadmium by interchanging the amino and carbamoyl groups on the fluorophore.
Tetrahydroindazolone substituted 2-aminobenzamides as fluorescent probes: switching metal ion selectivity from zinc to cadmium by interchanging the amino and carbamoyl groups on the fluorophore.
Organic & biomolecular chemistry (2012-06-23)
Jia Jia, Qin-Chao Xu, Ri-chen Li, Xi Tang, Ying-Fang He, Meng-Yu Zhang, Yuan Zhang, Guo-Wen Xing
RESUMEN
Three fluorescent probes CdABA', CdABA and ZnABA', which are structural isomers of ZnABA, have been designed with N,N-bis(2-pyridylmethyl) ethylenediamine (BPEA) as chelator and 2-aminobenzamide as fluorophore. These probes can be divided into two groups: CdABA, CdABA' for Cd(2+) and ZnABA, ZnABA' for Zn(2+). Although there is little difference in their chemical structures, the two groups of probes exhibit totally different fluorescence properties for preference of Zn(2+) or Cd(2+). In the group of Zn(2+) probes, ZnABA/ZnABA' distinguish Zn(2+) from Cd(2+) with F(Zn)(2+)-F(Cd)(2+) = 1.87-2.00. Upon interchanging the BPEA and carbamoyl groups on the aromatic ring of the fluorophore, the structures of ZnABA/ZnABA' are converted into CdABA/CdABA'. Interestingly, the metal ions selectivity of CdABA/CdABA' was switched to discriminate Cd(2+) from Zn(2+) with F(Cd)(2+)-F(Zn)(2+) = 2.27-2.36, indicating that a small structural modification could lead to a remarkable change of the metal ion selectivity. (1)H NMR titration and ESI mass experiments demonstrated that these fluorescent probers exhibited different coordination modes for Zn(2+) and Cd(2+). With CdABA' as an example, generally, upon addition of Cd(2+), the fluorescence response possesses PET pathway to display no obvious shift of maximum λ(em) in the absence or presence of Cd(2+). However, an ICT pathway could be employed after adding Zn(2+) into the CdABA' solution, resulting in a distinct red-shift of maximal λ(em).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anthranilamide, ≥98%