Merck
  • Regulation of the ERK pathway in the dentate gyrus by in vivo dopamine D1 receptor stimulation requires glutamatergic transmission.
Regulation of the ERK pathway in the dentate gyrus by in vivo dopamine D1 receptor stimulation requires glutamatergic transmission.
Neuropharmacology (2012-07-17)
Giuseppe Gangarossa, Emmanuel Valjent
RESUMEN
Acute systemic administration of the dopamine D1/D5 receptors (D1Rs) agonist, SKF81297, activates the extracellular signal-regulated protein kinases (ERK) pathway selectively in the granule cells of the dentate gyrus. In this study, we examined the mechanisms involved in this regulation and investigated the molecular components that could promote ERK-dependent transcription and translation. SKF81297 induced phosphorylation of ERK and histone H3 required intact glutamatergic transmission. Blockade of glutamate release achieved by the mGluR2/3 agonist, LY354740 or the selective adenosine A1R agonist, CCPA as well as neurotoxic lesions of lateral entorhinal cortex reduced the ability of SKF81297 to induce ERK activation in the dentate gyrus. This activation required the combined stimulation of NR2B-containing NMDARs, mGluR1 and mGluR5. SKF81297 evoked phosphorylation of the ribosomal protein S6 (rpS6) selectively at the Ser235/236 site while the Ser240/244 site remains unchanged. The SKF81297 induced increased phosphorylation of rpS6 was dependent on PKC and ERK/p90RSK activation. Surprisingly, administration of D1Rs agonist suppressed mTORC1/p70S6K pathway suggesting an mTOR-independent regulation of rpS6 phosphorylation. Taken together, our results show that intact glutamatergic transmission plays a major role in the regulation of ERK-dependent phosphorylation of histone H3 and rpS6 observed in the mouse dentate gyrus after systemic administration of SKF81297.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(±)-6-Chloro-PB hydrobromide, ≥98% (HPLC), solid