Merck
  • Influence of sutureless 23-gauge sclerotomy architecture on postoperative intraocular pressure decrease: results of a multivariate analysis.
Influence of sutureless 23-gauge sclerotomy architecture on postoperative intraocular pressure decrease: results of a multivariate analysis.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie (2012-10-31)
Rachid Tahiri Joutei Hassani, Mohamed El Sanharawi, Raphael Adam, Claire Monin, Sylvère Dupont-Monod, Christophe Baudouin
RESUMEN
To evaluate the factors affecting the postoperative intraocular pressure (IOP) decrease in 23-gauge (23-G) sutureless vitrectomy, including incision architecture evaluated by anterior segment spectral-domain optical coherence tomography (SD-OCT). A prospective cohort study of 43 patients who underwent primary transconjunctival 23-G pars plana vitrectomy. All sclerotomy wounds were imaged 1 day after surgery using the anterior segment module of SD-OCT (OCT Spectralis, Heidelberg Engineering, Heidelberg, Germany). 23-G sclerotomy architecture, preoperative and postoperative medical data were also prospectively collected. Multivariate logistic regression analysis, with backward elimination, found that surgery duration (adjusted OR = 9.17, p = 0.020) and loss of wound apposition (adjusted OR = 15.12, p = 0.022) were risk factors for significant postoperative IOP decrease (≥3 mmHg) 1 day after surgery; while age, gender, myopia, and gas tamponade were not risk or protective factors for postoperative IOP decrease. In 23-G pars plana vitrectomy, the early postoperative decrease in IOP is mainly influenced by surgery duration and the self-sealing nature of the sclerotomy. The IOP decrease was not influenced by the presence or the absence of gas tamponade.