Merck
First case of mirtazepine-induced Stevens-Johnson syndrome from India.
Indian journal of pharmacology (2012-11-01)
Amit Bhasin, Anil Gurtoo, Latika Gupta, Gunjan Aggarwal
RESUMEN
A 28-year-old woman, a known case of systemic lupus erythematosus (SLE), was admitted with mucocutaneous ulceroerosive lesions with blisters and thrombocytopenia after taking antidepressant mirtazepine. Exacerbation of SLE and drug-induced eruption was diagnosed. Clinical and laboratory markers were suggestive of Stevens-Johnson syndrome. This is a rare adverse effect of the newer generation antidepressant mirtazepine.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Mirtazapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Mianserin hydrochloride