Merck
Evaluation of in vivo anti-hyperglycemic and antioxidant potentials of α-santalol and sandalwood oil.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2013-02-02)
Biswapriya B Misra, Satyahari Dey
RESUMEN
Sandalwood finds numerous mentions across diverse traditional medicinal systems in use worldwide. The objective of this study was to evaluate the in vivo anti-hyperglycemic and antioxidant potential of sandalwood oil and its major constituent α-santalol. The in vivo anti-hyperglycemic experiment was conducted in alloxan-induced diabetic male Swiss albino mice models. The in vivo antioxidant experiment was performed in d-galactose mediated oxidative stress induced male Swiss albino mice models. Intraperitoneal administration of α-santalol (100mg/kg BW) and sandalwood oil (1g/kg BW) for an week modulated parameters such as body weight, blood glucose, serum bilirubin, liver glycogen, and lipid peroxides contents to normoglycemic levels in the alloxan-induced diabetic mice. Similarly, intraperitoneal administration of α-santalol (100mg/kg BW) and sandalwood oil (1g/kg BW) for two weeks modulated parameters such as serum aminotransferases, alkaline phosphatase, bilirubin, superoxide dismutase, catalase, free sulfhydryl, protein carbonyl, nitric oxide, liver lipid peroxide contents, and antioxidant capacity in d-galactose mediated oxidative stress induced mice. Besides, it was observed that the beneficial effects of α-santalol were well complimented, differentially by other constituents present in sandalwood oil, thus indicating synergism in biological activity of this traditionally used bioresource.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Sandalwood oil, natural