Merck
Dehydrogenative alkenylation of uracils via palladium-catalyzed regioselective C-H activation.
Chemical communications (Cambridge, England) (2013-03-27)
Yi-Yun Yu, Gunda I Georg
RESUMEN
A regioselective Pd-catalyzed cross-dehydrogenative coupling between uracils and alkenes is reported. This protocol provides easy access to a variety of 5-alkenyluracil structural motifs.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture