Merck
[Achilles heel of COPD].
Praxis (2013-07-05)
Stefanie Schuster, Markus Diethelm
RESUMEN
We report the case of a 73 year old patient with a COPD. A severe exazerbation was treated with glucocorticoids and Quinolones. A few days later both ankles were tender and painful. Ultrasound examination showed bilateral Achilles tendon ruptures. Diagnostics, etiology and therapeutic options are discussed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Prednisona, ≥98%
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Prednisona, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Levofloxacin, analytical standard
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ofloxacin, VETRANAL®, analytical standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard