Merck
  • [Characteristics of dimedrol, pipolfen and suprastin absorption from the small intestine of rats in systemic anaphylactic shock].
[Characteristics of dimedrol, pipolfen and suprastin absorption from the small intestine of rats in systemic anaphylactic shock].
Farmakologiia i toksikologiia (1988-03-01)
K I Bender, A N Lutsevich, I N Lutsevich
RESUMEN
It was shown by the in situ method that absorption of dimedrol, pipolphen and suprastine from the small intestine of the rats immunized parenterally with ovalbumin against the background of intravenous administration of the resolving dose of ovalbumin significantly decreased in connection with the formation in the immunized rats of antiovalbumin antibodies. Administration of euphylline simultaneously with the resolving dose of ovalbumin to the immunized rats partially normalized absorbability of all three drugs.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Chloropyramine hydrochloride, analytical standard, for drug analysis
En este momento no podemos mostrarle ni los precios ni la disponibilidad