Merck
[Separation of [Rh-103m]-rhodocene derivatives from the parent [103Ru]ruthenocene derivatives and their organ distribution].
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes (1987-01-01)
M Wenzel, Y F Wu
RESUMEN
The radioactive decay of [103Ru]ruthenocene derivatives leads to 103mRh labelled rhodocinium derivatives, which can be separated by the extraction of a lipophilic solution of the ruthenocen derivate with water. The separation factor 103mRh/103Ru reaches values of 32:1 Rh3+ ions are not liberated and extracted. The organ distribution of the 103mRh labelled rhodocinium derivatives gained from ruthenocene and from N-isopropyl-ruthenocene amphetamine is different from the distribution of the parent ruthenocene compound. The liver and kidney uptake of the rhodocinium-amphetamine is much higher than the uptake with ruthenocene amphetamine.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bis(cyclopentadienyl)ruthenium(II), 97%