Merck
HPr/HPr-P phosphoryl exchange reaction catalyzed by the mannitol specific enzyme II of the bacterial phosphotransferase system.
The Journal of biological chemistry (1987-02-25)
S L Sutrina, E B Waygood, F C Grenier, M H Saier
RESUMEN
The mannitol specific Enzyme II of the phosphoenolpyruvate: sugar phosphotransferase system of Escherichia coli catalyzes an exchange reaction in which a phosphoryl moiety is transferred from one molecule of the heat stable phosphocarrier protein HPr to another. An assay was developed for measuring this reaction. Unlabeled phospho-HPr and 125I-labeled free HPr were incubated together in the presence of Enzyme IImtl, and production of 125I-labeled phospho-HPr was measured. The reaction was concentration-dependent with respect to Enzyme IImtl and did not occur in its absence. The reaction occurred in the absence of Mg2+ in the presence of 10 mM EDTA. Treatment of Enzyme IImtl with the histidyl reagent diethylpyrocarbonate inactivated it with respect to the exchange reaction. Levels of N-ethylmaleimide which inactivate Enzyme IImtl with respect to both P-enolpyruvate-dependent phosphorylation of mannitol and mannitol/mannitol-1-P transphosphorylation did not affect its activity in the exchange reaction; however, treatment with another sulfhydryl reagent, p-chloromercuribenzoate, resulted in partial inactivation. The pH optimum for the Enzyme IImtl-catalyzed exchange reaction was about 7.5. Enzyme I and the glucose specific Enzyme III, two other E. coli phosphotransferase system proteins which, like Enzyme IImtl, interact directly with HPr, were also shown to catalyze 125I-HPr/HPr-P phosphoryl exchange.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
D-Mannitol 1-phosphate lithium salt, ≥95% (TLC)