Merck
[Enzymatic hydroxylation of ruthenocene and osmocene by liver microsomes].
Zeitschrift fur Naturforschung. Section C, Biosciences (1982-01-01)
M Schneider, M Wenzel, G Schachschneider
RESUMEN
In buffer solution (pH 7,4) ruthenocene and osmocene are far more stable than ferrocene. The metallocenes ruthenocene and osmocene are metabolized by microsomes of mouse liver in the presence of NADPH and O2. The Km-values are similar for both metallocenes, the rate of metabolism is higher for osmocene that for ruthenocene.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bis(cyclopentadienyl)ruthenium(II), 97%