- Biotransformation of benzyl thiocyanate by Streptomyces aureofaciens.
Biotransformation of benzyl thiocyanate by Streptomyces aureofaciens.
Letters in applied microbiology (1994-09-01)
J Fuska, M Sturdíkovă, B Proksa
PMID7765389
RESUMEN
Benzyl thiocyanate, a specific stimulator of chlortetracycline biosynthesis, was transformed into dibenzyl disulphide by Streptomyces aureofaciens. The disulphide stimulated chlortetracycline production to a lesser extent than did benzyl thiocyanate.
MATERIALES