Merck
Alternatives to arginine as energy sources for the non-fermentative Mycoplasma gallinarum.
FEMS microbiology letters (1994-01-15)
R R Taylor, H Varsani, R J Miles
RESUMEN
In contrast to previously studied non-fermentative arginine-hydrolysing (F-/A+) Mycoplasma species, M. gallinarum cells suspended in a salts solution oxidised ethanol and L-lactic, pyruvic and 2-oxobutyric acids. The organic acids were additionally shown effectively to replace arginine as energy sources in growth media. However, their presence did not inhibit arginine hydrolysis, nor did arginine inhibit organic acid catabolism. The ability to oxidise organic acids is a potentially useful diagnostic character enabling sub-division of the F-/A+ Mycoplasma species. M. gallinarum also differed from previously studied F-/A+ mycoplasmas in possessing relatively high NADH oxidase activity and producing H2O2 as only a minor product of NADH oxidation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
β-Fluoropyruvic acid sodium salt monohydrate, ≥98%