Merck
  • Urinary screening for adinazolam and its major metabolites by the Emit d.a.u. and FPIA benzodiazepine assays with confirmation by HPLC.
Urinary screening for adinazolam and its major metabolites by the Emit d.a.u. and FPIA benzodiazepine assays with confirmation by HPLC.
Journal of analytical toxicology (1993-11-01)
A D Fraser, A F Isner, W Bryan
RESUMEN
Adinazolam is a triazolobenzodiazepine, currently under clinical investigation, that possesses antidepressant and anxiolytic activity. It has a short half-life (less than 3 h), and less than 2% of an oral dose is excreted unchanged. The major urinary metabolite is N-desmethyladinazolam, and minor metabolites are estazolam and alpha-OH-alprazolam. The objective of this study was to characterize the reactivity of adinazolam, N-desmethyladinazolam, and estazolam in the Emit d.a.u. benzodiazepine assay and the Abbott TDx urine (FPIA) benzodiazepine assay. N-desmethyladinazolam and estazolam gave an equivalent response to the Emit cutoff calibrator (300 ng/mL) at 100-200 ng/mL, and adinazolam gave an equivalent response at 200 ng/mL. By FPIA, N-desmethyladinazolam and adinazolam had equivalent net polarization values as the 300-ng/mL low control at 500-1000 ng/mL, and estazolam gave a positive response at 300 ng/mL. Six volunteers received single oral doses of 10, 30, and 50 mg of adinazolam. Urine specimens (N = 7) were collected from 0 to 36 h post-administration. By Emit, all urine specimens at all doses were positive from 2 to 36 h, and all FPIA analyzed specimens were positive from 2 to 24 h. Confirmation testing was performed by HPLC by analyzing for N-desmethyladinazolam. All urine specimens were confirmed positive for N-desmethyladinazolam (greater than 200 ng/mL) except for the blank specimens (time = 0) and 7 of 18 specimens collected 36 h post-administration. In conclusion, both immunoassay screening assays are acceptable for detecting the presence of adinazolam in human urine for up to 24 h after a single oral dose of 10-50 mg.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
alpha-Hydroxyalprazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
α-Hydroxyalprazolam, powder, ≥98% (TLC)