Merck
Synthesis and SAR of bulky 1-azabicyclo[2.2.1]-3-one oximes as muscarinic receptor subtype selective agonists.
Life sciences (1993-01-01)
H Tecle, D J Lauffer, T Mirzadegan, W H Moos, D W Moreland, M R Pavia, R D Schwarz, R E Davis
RESUMEN
The synthesis of a series of potent and efficacious 1-azabicyclo[2.2.1]heptan-3-one oxime muscarinic agonists is described. The oximes have extended appendages designed to span the cavity defined by the seven transmembrane helices of the muscarinic receptor. Some members of the series are selective for receptors of the m1 subtype. One such oxime, 31, shows affinity and functional selectivity for m1 over m2, m3, and m4 muscarinic receptor types.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2-Methyl-1,3-dioxolane, 97%