Merck
Anti-arthritic effects of the novel dipeptidyl peptidase IV inhibitors TMC-2A and TSL-225.
Immunopharmacology (1998-10-17)
S Tanaka, T Murakami, N Nonaka, T Ohnuki, M Yamada, T Sugita
RESUMEN
We evaluated the immunopharmacological effects of two novel dipeptidyl peptidase IV (DP IV) inhibitors, TMC-2A [(2S,2S',2S'')-2-[2'-[2''-amino-3''-(-indol-3'''-yl)-1''-oxopropyl]-1',2 ',3',4'-tetrahydro-6',8'-dihydroxy-7'-methoxyisoquinol-3-yl-car bonylamino]-4-hydroxymethyl-5-hydroxypentanoic acid] and TSL-225 (tryptophyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid). TMC-2A, produced by Aspergillus sp. A374, inhibited rat kidney DP IV uncompetitively, with a Ki value of 5.3 microM. In vivo, TMC-2A suppressed alkyldiamine (N,N-dioctadecyl-N',N-bis(2-hydroxyethyl)propanediamine)-induced arthritis. We developed a chemically modified inhibitor, TSL-225, with potency similar to that of TMC-2A. TSL-225 inhibited DP IV uncompetitively, with a Ki value of 3.6 microM. TSL-225 was also effective against adjuvant-induced arthritis. These results suggest that TMC-2A and its derivatives may have therapeutic potential for the treatment of inflammatory diseases such as rheumatoid arthritis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Avridine, ≥97% (HPLC)