Merck
Blockade of C5a and C5b-9 generation inhibits leukocyte and platelet activation during extracorporeal circulation.
The Journal of clinical investigation (1995-09-01)
C S Rinder, H M Rinder, B R Smith, J C Fitch, M J Smith, J B Tracey, L A Matis, S P Squinto, S A Rollins
RESUMEN
Complement activation contributes to the systemic inflammatory response induced by cardiopulmonary bypass. At the cellular level, cardiopulmonary bypass activates leukocytes and platelets; however the contribution of early (3a) versus late (C5a, soluble C5b-9) complement components to this activation is unclear. We used a model of simulated extracorporeal circulation that activates complement (C3a, C5a, and C5b-9 formation), platelets (increased percentages of P-selectin-positive platelets and leukocyte-platelet conjugates), and neutrophils (upregulated CD11b expression). to specifically target complement activation in this model, we added a blocking mAb directed at the human C5 complement component and assessed its effect on complement and cellular activation. Compared with a control mAB, the anti-human C5 mAb profoundly inhibited C5a and soluble C5b-9 generation and serum complement hemolytic activity but had no effect on C3a generation. Additionally, the anti-human C5 mAb significantly inhibited neutrophil CD11b upregulation and abolished the increase in P-selectin-positive platelets and leukocyte-platelet conjugate formation compared to experiments performed with the control mAb. This suggests that the terminal components C5a and C5b-9, but not C3a, directly contribute to platelet and neutrophil activation during extracorporeal circulation. Furthermore, these data identify the C5 component as a site for therapeutic intervention in cardiopulmonary bypass.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium 5,5-diethylbarbiturate, purum, ≥99.0% (NT)