Merck
  • Effects of transcranial focal electrical stimulation alone and associated with a sub-effective dose of diazepam on pilocarpine-induced status epilepticus and subsequent neuronal damage in rats.
Effects of transcranial focal electrical stimulation alone and associated with a sub-effective dose of diazepam on pilocarpine-induced status epilepticus and subsequent neuronal damage in rats.
Epilepsy & behavior : E&B (2013-07-28)
Walter Besio, Manola Cuellar-Herrera, Hiram Luna-Munguia, Sandra Orozco-Suárez, Luisa Rocha
RESUMEN
Experiments were conducted to evaluate the effects of transcranial focal electrical stimulation (TFS) applied via tripolar concentric ring electrodes, alone and associated with a sub-effective dose of diazepam (DZP) on the expression of status epilepticus (SE) induced by lithium-pilocarpine (LP) and subsequent neuronal damage in the hippocampus. Immediately before pilocarpine injection, male Wistar rats received TFS (300Hz, 200-μs biphasic square charge-balanced 50-mA constant current pulses for 2min) alone or combined with a sub-effective dose of DZP (0.41mg/kg, i.p.). In contrast with DZP or TFS alone, DZP plus TFS reduced the incidence of, and enhanced the latency to, mild and severe generalized seizures and SE induced by LP. These effects were associated with a significant reduction in the number of degenerated neurons in the hippocampus. The present study supports the notion that TFS combined with sub-effective doses of DZP may represent a therapeutic tool to induce anticonvulsant effects and reduce the SE-induced neuronal damage.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cloruro de litio, ACS reagent, ≥99%
Sigma-Aldrich
Cloruro de litio, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Cloruro de litio, Molecular Biology, ≥99%
Sigma-Aldrich
Cloruro de litio, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Diazepam
Sigma-Aldrich
Pilocarpine hydrochloride, ≥99% (titration), powder
Supelco
Lithium chloride solution, 1 M in ethanol
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Cloruro de litio, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Cloruro de litio, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Pilocarpine nitrate salt, ≥98% (HPLC)
Sigma-Aldrich
Cloruro de litio, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Cloruro de litio, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pilocarpine nitrate, meets USP testing specifications
Pilocarpine nitrate for system suitability, European Pharmacopoeia (EP) Reference Standard