Merck
  • Synthesis of a tetrasubstituted tetrahydronaphthalene scaffold for α-helix mimicry via a MgBr2-catalyzed Friedel-Crafts epoxide cycloalkylation.
Synthesis of a tetrasubstituted tetrahydronaphthalene scaffold for α-helix mimicry via a MgBr2-catalyzed Friedel-Crafts epoxide cycloalkylation.
Organic letters (2013-09-11)
Devan Naduthambi, Santosh Bhor, Michael B Elbaum, Neal J Zondlo
RESUMEN
α-Helices are ubiquitous protein recognition elements that bind diverse biomolecular targets. The synthesis of a small molecule scaffold to present the side chains of an α-helix is described. The 1,3,5,7-tetrasubstituted 1,2,3,4-tetrahydronaphthalene scaffold, providing mimicry of the i, i+3, and i+4 positions of an α-helix, was synthesized using a novel MgBr2-catalyzed Friedel-Crafts epoxide cycloalkylation as the key step. Each position may be differentiated via O-alkylation after scaffold synthesis, generating a diversity-oriented approach to readily synthesize proteomimetics for different targets.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Melting point standard 79.5-81°C, analytical standard
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland