Merck
  • Novel pH-sensitive micelles generated by star-shape copolymers containing zwitterionic sulfobetaine for efficient cellular internalization.
Novel pH-sensitive micelles generated by star-shape copolymers containing zwitterionic sulfobetaine for efficient cellular internalization.
Journal of biomedical nanotechnology (2013-09-26)
Jun Cao, Shuying Zhai, Chenlong Li, Bin He, Yusi Lai, Yuanwei Chen, Xianglin Luo, Zhongwei Gu
RESUMEN
pH-sensitive micelles are considered promising carriers for tumor targeted drug delivery. In this study, novel pH-sensitive star-shape copolymers of amphiphilic poly(epsilon-caprolactone)-b-poly(N, N-diethylaminoethyl methacrylate)-r-poly(N-(3-sulfopropyl)-N-methacryloxyethy-N, N-diethylammoniumbetaine) (4sPCLDEAS) are designed and synthesized with the combination of ring opening polymerization (ROP) and atom radical transferpolymerization (ATRP). The structure of the copolymers is characterized by proton nuclear magnetic resonance spectra (1HNMR). The poly(N-(3-sulfopropyl)-N-methacryloxyethy-N, N-diethylammoniumbetaine) segment is used instead of poly(ethylene glycol) (PEG) as hydrophilic block in the copolymers to form polymeric micelles. The micelles present spherical shape, narrow size distribution, and are reponsive to the acidity. The CMC of the micelles is as low as 1 x 10(-3) mg mL(-1). Doxorubin (DOX) is efficiently encapsulated in the micelles and the drug release is pH dependant. The cytotoxicity as well as the intracellular drug delivery of the micelles are investigated. The micelles are nontoxic to human cervical carcinoma (Hela) cells. The DOX-loaded micelles are internalized in Hela cells efficiently, which are better than that of hydrophilic doxorubicin hydrochloride (DOX x HCl). These pH-sensitive micelles are potential promising carriers for anti-cancer drug delivery.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Betaína solution, 5 M, PCR Reagent
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Betaine hydrochloride, ≥99%