Merck
Cyclic 100-ka (glacial-interglacial) migration of subseafloor redox zonation on the Peruvian shelf.
Proceedings of the National Academy of Sciences of the United States of America (2013-10-23)
Sergio Contreras, Patrick Meister, Bo Liu, Xavier Prieto-Mollar, Kai-Uwe Hinrichs, Arzhang Khalili, Timothy G Ferdelman, Marcel M M Kuypers, Bo Barker Jørgensen
RESUMEN
The coupling of subseafloor microbial life to oceanographic and atmospheric conditions is poorly understood. We examined diagenetic imprints and lipid biomarkers of past subseafloor microbial activity to evaluate its response to glacial-interglacial cycles in a sedimentary section drilled on the Peruvian shelf (Ocean Drilling Program Leg 201, Site 1229). Multiple and distinct layers of diagenetic barite and dolomite, i.e., minerals that typically form at the sulfate-methane transition (SMT), occur at much shallower burial depth than the present SMT around 30 meters below seafloor. These shallow layers co-occur with peaks of (13)C-depleted archaeol, a molecular fossil of anaerobic methane-oxidizing Archaea. Present-day, non-steady state distributions of dissolved sulfate also suggest that the SMT is highly sensitive to variations in organic carbon flux to the surface shelf sediments that may lead to shoaling of the SMT. Reaction-transport modeling substantiates our hypothesis that shallow SMTs occur in response to cyclic sediment deposition with a high organic carbon flux during interglacials and a low organic carbon flux during glacial stages. Long diffusion distances expectedly dampen the response of deeply buried microbial communities to changes in sediment deposition and other oceanographic drivers over relatively short geological time scales, e.g., glacial-interglacial periods. However, our study demonstrates how dynamically sediment biogeochemistry of the Peru Margin has responded to glacial-interglacial change and how these changes are now preserved in the geological record. Such changes in subsurface biogeochemical zonation need to be taken into account to assess the role of the subseafloor biosphere in global element and redox cycling.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Carbonato de calcio, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Barium sulfate, ReagentPlus®, 99%
Sigma-Aldrich
Carbonato de calcio, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Magnesio, powder, ≥99%
Sigma-Aldrich
Magnesio, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesio, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesio, turnings, reagent grade, 98%
Sigma-Aldrich
Carbonato de calcio, ReagentPlus®
Sigma-Aldrich
Carbonato de calcio, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Carbonato de calcio, ACS reagent, 99.95-100.05% dry basis
Supelco
Carbonato de calcio, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Magnesio, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Barium sulfate, 99.99% trace metals basis
Sigma-Aldrich
Carbonato de calcio, 99.999% trace metals basis
Sigma-Aldrich
Magnesio, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Carbonato de calcio, ≥99.995% trace metals basis
Sigma-Aldrich
Carbonato de calcio, BioXtra, ≥99.0%
Sigma-Aldrich
Carbonato de calcio, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Carbonato de calcio, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Barium sulfate, meets USP testing specifications
Sigma-Aldrich
Magnesio, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesio, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesio, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Carbonato de calcio, tested according to Ph. Eur.
Sigma-Aldrich
Magnesio, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesio, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesio, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesio, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesio, in a Sure/Seal bottle, turnings, 37.5 mmol