Merck
  • Synthesis, characterization, and antimicrobial evaluation of a small library of ferrocene-containing acetoacetates and phenyl analogs: the discovery of a potent anticandidal agent.
Synthesis, characterization, and antimicrobial evaluation of a small library of ferrocene-containing acetoacetates and phenyl analogs: the discovery of a potent anticandidal agent.
Molecular diversity (2014-03-14)
Niko S Radulović, Marko Z Mladenović, Zorica Stojanović-Radić, Goran A Bogdanović, Dragana Stevanović, Rastko D Vukićević
RESUMEN
A library of 16 2-substituted methyl acetoacetates containing ferrocenyl or phenyl units was designed to disclose differences in the antimicrobial activity of ferrocene-containing compounds and their phenyl analogs. Two methyl acetoacetates, whose structures do not contain an aromatic nucleus, were also included in order to probe the inherent activity of the scaffold itself. The acetoacetates were synthesized (low-to-good yields) and fully characterized by spectral (MS, IR, UV-Vis, 1D and 2D NMR) and electrochemical (cyclic voltammetry) techniques. Single-crystal X-ray analysis has been performed for methyl 2-acetyl-2-(ferrocenylmethyl)-5-methylhex-4-enoate. All compounds have demonstrated in vitro antimicrobial activity against six bacterial (three Gram-positive and three Gram-negative) and two fungal strains with minimal inhibitory concentration values of 0.0050-20.6 μmol mL(-1). The most active compound was 2-acetyl-2-(ferrocenylmethyl)-4-methylpent-4-enoate whose activity was comparable to that of nystatin against the yeast Candida albicans. Agglomerative hierarchical clustering statistical analysis of the antimicrobial assay data demonstrated that ferrocene-containing compounds have statistically different and greater antimicrobial activity when compared to their phenyl analogs.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
GW833972A, ≥98% (HPLC), powder