Merck
  • Secreted stress-induced phosphoprotein 1 activates the ALK2-SMAD signaling pathways and promotes cell proliferation of ovarian cancer cells.
Secreted stress-induced phosphoprotein 1 activates the ALK2-SMAD signaling pathways and promotes cell proliferation of ovarian cancer cells.
Cell reports (2012-08-14)
Chia-Lung Tsai, Chi-Neu Tsai, Chiao-Yun Lin, Hsi-Wen Chen, Yun-Shien Lee, Angel Chao, Tzu-Hao Wang, Hsin-Shih Wang, Chyong-Huey Lai
RESUMEN
Stress-induced phosphoprotein 1 (STIP1), a cochaperone that organizes other chaperones, heat shock proteins (HSPs), was recently shown to be secreted by human ovarian cancer cells. In neuronal tissues, binding to prion protein was required for STIP1 to activate the ERK (extracellular-regulated MAP kinase) signaling pathways. However, we report that STIP1 binding to a bone morphogenetic protein (BMP) receptor, ALK2 (activin A receptor, type II-like kinase 2), was necessary and sufficient to stimulate proliferation of ovarian cancer cells. The binding of STIP1 to ALK2 activated the SMAD signaling pathway, leading to transcriptional activation of ID3 (inhibitor of DNA binding 3), promoting cell proliferation. In conclusion, ovarian-cancer-tissue-secreted STIP1 stimulates cancer cell proliferation by binding to ALK2 and activating the SMAD-ID3 signaling pathways. Although animal studies are needed to confirm these mechanisms in vivo, our results may pave the way for developing novel therapeutic strategies for ovarian cancer.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-STIP1 (AB1) antibody produced in rabbit, IgG fraction of antiserum