Merck
Fish odor syndrome: a case report of trimethylaminuria.
Dermatology online journal (2014-01-25)
Catherine A Ulman, Julian J Trevino, Marvin Miller, Rishi K Gandhi
RESUMEN
Trimethylaminuria is a rare, autosomal recessive, metabolic disorder that results in accumulation of trimethylamine (TMA), which smells like rotten fish. The chemical is excreted in sweat and urine owing to a deficiency in the enzyme flavin monooxygenase 3 (FMO3). We report a case of trimethylaminuria in a 12-year-old girl. The patient failed treatment with diet and hygiene modification, but achieved symptomatic improvement after a four-month course of metronidazole.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-FMO3 antibody produced in rabbit, affinity isolated antibody