Merck
Mutations in FAM134B, encoding a newly identified Golgi protein, cause severe sensory and autonomic neuropathy.
Nature genetics (2009-10-20)
Ingo Kurth, Torsten Pamminger, J Christopher Hennings, Désirée Soehendra, Antje K Huebner, Annelies Rotthier, Jonathan Baets, Jan Senderek, Haluk Topaloglu, Sandra A Farrell, Gudrun Nürnberg, Peter Nürnberg, Peter De Jonghe, Andreas Gal, Christoph Kaether, Vincent Timmerman, Christian A Hübner
RESUMEN
Hereditary sensory and autonomic neuropathy type II (HSAN II) leads to severe mutilations because of impaired nociception and autonomic dysfunction. Here we show that loss-of-function mutations in FAM134B, encoding a newly identified cis-Golgi protein, cause HSAN II. Fam134b knockdown results in structural alterations of the cis-Golgi compartment and induces apoptosis in some primary dorsal root ganglion neurons. This implicates FAM134B as critical in long-term survival of nociceptive and autonomic ganglion neurons.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-FAM134B (lumenal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-FAM134B antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution