Merck
Glycosidic fraction of flue-cured tobacco leaves: its separation and component analysis.
Bioscience, biotechnology, and biochemistry (2000-05-10)
K Ito, Y Tanabe, S Kato, T Yamamoto, A Saito, M Mori
RESUMEN
The fraction containing glycosidic components was separated from flue-cured tobacco (Nicotiana tabacum L.) leaves by a facile method. Some components of the fraction were isolated and elucidated to be syringin, coniferin, cichoriin, benzyl-beta-D-glucoside, Blumenol A-beta-D-glucoside, and 5,6-epoxy-5,6-dihydro-3-hydroxy-beta-ionyl-beta-D-glucoside. Syringin and coniferin were detected in the Nicotiana species for the first time.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Abietin, ≥90% (LC/MS-ELSD)