Merck
  • Quantification of BNP7787 (dimesna) and its metabolite mesna in human plasma and urine by high-performance liquid chromatography with electrochemical detection.
Quantification of BNP7787 (dimesna) and its metabolite mesna in human plasma and urine by high-performance liquid chromatography with electrochemical detection.
Journal of chromatography. B, Biomedical sciences and applications (2001-05-04)
M Verschraagen, T H Zwiers, P E de Koning, J Welink, W J van der Vijgh
RESUMEN
A sensitive and accurate assay was developed and validated to determine BNP7787 (dimesna), a new protector against cisplatin-induced toxicities, and its metabolite mesna in plasma and urine of patients. Both analytes were measured as mesna in deproteinized plasma or in urine diluted with mobile phase using high-performance liquid chromatography with an electrochemical detector provided with a wall-jet gold electrode. The assays for BNP7787 and mesna in deproteinized plasma were linear over the range of 1.6-500 microM and 0.63-320 microM, respectively. In plasma, the mean recovery of BNP7787 over the whole concentration range was 100.6% and of mesna 94.6%. The lower limits of quantitation (LLQs) of BNP7787 and mesna in deproteinized plasma were 1.6 microM and 0.63 microM, respectively. For both compounds the within- and between-day accuracy and precision of the assay was better than 12%. The assays for BNP7787 and mesna in urine were linear over the range of 0.8-1200 microM and 0.63-250 microM, respectively. In urine, the mean recovery of BNP7787 over the whole concentration range was 94.1% and of mesna 93.1%. The LLQ of BNP7787 in urine was 0.8 microM and of mesna 1.6 microM. The within- and between-day accuracy and precision of the assay for BNP7787 and mesna was lower than 15%. The stability of mesna in urine increased with an increasing concentration of mesna, lower temperature and addition of EDTA (1 g/l) and hydrochloric acid (0.2 M). BNP7787 in urine was stable for at least 24 h at temperatures in the range of -20 degrees C up to 37 degrees C and independent of the concentration. The developed assays are currently applied for samples of patients with solid tumors participating in a phase I trial of BNP7787 in combination with cisplatin.
MATERIALES
Número de producto
Marca
Descripción del producto
Mesna impurity D, European Pharmacopoeia (EP) Reference Standard