Merck
  • Use of near-infrared reflection spectroscopy to study the effects of X-ray contrast media on renal tolerance in rats: effects of a prostacyclin analogue and of phosphodiesterase inhibitors.
Use of near-infrared reflection spectroscopy to study the effects of X-ray contrast media on renal tolerance in rats: effects of a prostacyclin analogue and of phosphodiesterase inhibitors.
Investigative radiology (2002-11-26)
W Krause, P Muschick, U Krüger
RESUMEN
Use of near-infrared reflection spectroscopy (NIR-RS) as a new model to assess renal tolerance of contrast agents and determination of the effects of a prostacyclin analogue and of two phosphodiesterase inhibitors on renal tolerance. NIR-RS was used to measure total hemoglobin, oxygenated hemoglobin and tissue oxygen saturation in the renal cortex of rats and the effect of diatrizoate, iopromide and iotrolan injected at 1 g iodine/kg alone or together with the prostacyclin derivative, iloprost, or the phosphodiesterase inhibitors, rolipram and mesopram, on these parameters. Injection of the contrast media alone resulted in a 10% to 35% depression of total hemoglobin, oxygenated hemoglobin, and tissue oxygen saturation approximately 40 to 100 seconds after administration, whereas saline showed no effect and mannitol solution only a minor effect. Coadministration of iloprost or pretreatment with the phosphodiesterase inhibitors, rolipram or mesopram, significantly attenuated the contrast media-induced effects. NIR-RS might be useful for the determination of contrast media-induced side effects. Stable prostacyclin analogues or phosphodiesterase inhibitors have the potential to mitigate these side effects.
MATERIALES
Número de producto
Marca
Descripción del producto
Iotrolan for system suitability, European Pharmacopoeia (EP) Reference Standard
Iotrolan, European Pharmacopoeia (EP) Reference Standard