Merck
Determination of dimethindene enantiomers in pharmaceuticals by capillary electrophoresis with carboxyethyl-beta-cyclodextrin.
Die Pharmazie (2007-02-14)
P Mikus, P Kubacák, I Valásková, E Havránek
RESUMEN
Cyclodextrin-mediated capillary zone electrophoresis in a hydrodynamically closed separation system with suppressed electroosmotic flow and UV absorbance photometric detection was developed for a high effective separation and quantitation of dimethindene enantiomers in various pharmaceutical formulations (solution, gel, capsules). The running buffer consisted of epsilon-aminocaproic acid (20 mmol/l) adjusted to pH 4.5 with acetic acid, negatively charged carboxyethyl-beta-cyclodextrin (2.5 mg/ml), serving as chiral selector, and 0.1% (w/v) methylhydroxyethylcellulose, serving as an electroosmotic flow suppressor. The proposed method was successfully validated appraising parameters of sensitivity, linearity, precision, accuracy/recovery and robustness and it is useful for routine use.
MATERIALES
Número de producto
Marca
Descripción del producto
Dimetindene maleate, European Pharmacopoeia (EP) Reference Standard