Merck
  • Overexpression of phosphoserine aminotransferase PSAT1 stimulates cell growth and increases chemoresistance of colon cancer cells.
Overexpression of phosphoserine aminotransferase PSAT1 stimulates cell growth and increases chemoresistance of colon cancer cells.
Molecular cancer (2008-01-29)
Nadia Vié, Virginie Copois, Caroline Bascoul-Mollevi, Vincent Denis, Nicole Bec, Bruno Robert, Caroline Fraslon, Emmanuel Conseiller, Franck Molina, Christian Larroque, Pierre Martineau, Maguy Del Rio, Céline Gongora
RESUMEN
Colorectal cancer (CRC) is one of the most common causes of cancer death throughout the world. In this work our aim was to study the role of the phosphoserine aminotransferase PSAT1 in colorectal cancer development. We first observed that PSAT1 is overexpressed in colon tumors. In addition, we showed that after drug treatment, PSAT1 expression level in hepatic metastases increased in non responder and decreased in responder patients. In experiments using human cell lines, we showed that ectopic PSAT1 overexpression in colon carcinoma SW480 cell line resulted in an increase in its growth rate and survival. In addition, SW480-PSAT1 cells presented a higher tumorigenic potential than SW480 control cells in xenografted mice. Moreover, the SW480-PSAT1 cell line was more resistant to oxaliplatin treatment than the non-transfected SW480 cell line. This resistance resulted from a decrease in the apoptotic response and in the mitotic catastrophes induced by the drug treatment. These results show that an enzyme playing a role in the L-serine biosynthesis could be implicated in colon cancer progression and chemoresistance and indicate that PSAT1 represents a new interesting target for CRC therapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human PSAT1