Merck
Cooperation of factor VII-activating protease and serum DNase I in the release of nucleosomes from necrotic cells.
Arthritis & rheumatology (Hoboken, N.J.) (2014-02-28)
Femke Stephan, Gerben Marsman, Liza M Bakker, Ingrid Bulder, Fabian Stavenuiter, Lucien A Aarden, Sacha Zeerleder
RESUMEN
Removal of dead cells is essential in the maintenance of tissue homeostasis, and efficient removal prevents exposure of intracellular content to the immune system, which could lead to autoimmunity. The plasma protease factor VII-activating protease (FSAP) can release nucleosomes from late apoptotic cells. FSAP circulates as an inactive single-chain protein, which is activated upon contact with either apoptotic cells or necrotic cells. The purpose of this study was to investigate the role of FSAP in the release of nucleosomes from necrotic cells. Necrotic Jurkat cells were incubated with serum, purified 2-chain FSAP, and/or DNase I. Nucleosome release was analyzed by flow cytometry, and agarose gel electrophoresis was performed to detect DNA breakdown. Incubation with serum released nucleosomes from necrotic cells. Incubation with FSAP-deficient serum or serum in which FSAP was inhibited by a blocking antibody was unable to release nucleosomes from necrotic cells, confirming that FSAP is indeed the essential serum factor in this process. Together with serum DNase I, FSAP induced the release of DNA from the cells, the appearance of nucleosomes in the supernatant, and the fragmentation of chromatin into eventually mononucleosomes. FSAP and DNase I are the essential serum factors that cooperate in necrotic cell DNA degradation and nucleosome release. We propose that this mechanism may be important in the removal of potential autoantigens.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Desoxirribonucleasa I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Desoxirribonucleasa I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Desoxirribonucleasa I from bovine pancreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Desoxirribonucleasa I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Desoxirribonucleasa I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
DNase I, Amplification Grade
Sigma-Aldrich
Deoxyribonuclease I RNase-free solution from bovine pancreas
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free