Merck
Phylogeny poorly predicts the utility of a challenging horizontally transferred gene in Methylobacterium strains.
Journal of bacteriology (2014-04-01)
Joshua K Michener, Stéphane Vuilleumier, Françoise Bringel, Christopher J Marx
RESUMEN
Horizontal gene transfer plays a crucial role in microbial evolution. While much is known about the mechanisms that determine whether physical DNA can be transferred into a new host, the factors determining the utility of the transferred genes are less clear. We have explored this issue using dichloromethane consumption in Methylobacterium strains. Methylobacterium extorquens DM4 expresses a dichloromethane dehalogenase (DcmA) that has been acquired through horizontal gene transfer and allows the strain to grow on dichloromethane as the sole carbon and energy source. We transferred the dcmA gene into six Methylobacterium strains that include both close and distant evolutionary relatives. The transconjugants varied in their ability to grow on dichloromethane, but their fitness on dichloromethane did not correlate with the phylogeny of the parental strains or with any single tested physiological factor. This work highlights an important limiting factor in horizontal gene transfer, namely, the capacity of the recipient strain to accommodate the stress and metabolic disruption resulting from the acquisition of a new enzyme or pathway. Understanding these limitations may help to rationalize historical examples of horizontal transfer and aid deliberate genetic transfers in biotechnology for metabolic engineering.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Tributyltin hydride, contains 0.05% BHT as stabilizer, 97%
Supelco
Diclorometano, analytical standard
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
[Arg8]-Vasopressin acetate salt, ≥95% (HPLC)
Sigma-Aldrich
Diclorometano, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
[Arg8]-Vasopressin solution, Grade VI (synthetic), ~100 IU/mL in 0.9% NaCl
Supelco
Diclorometano, Selectophore, ≥99.5%
Sigma-Aldrich
[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11, ≥97% (HPLC)