Merck
Nateglinide (Starlix): update on a new antidiabetic agent.
International journal of clinical practice (2003-08-16)
L S Phillips, B E Dunning
RESUMEN
Nateglinide is a new oral antidiabetic agent that stimulates insulin release promptly after its pre-meal administration in a strongly glucose-dependent fashion. Because its insulinotropic effects are short in duration, nateglinide specifically targets postprandial hyperglycaemia with a low potential to elicit hypoglycaemia or sustained hyperinsulinaemia. Nateglinide has an excellent safety and tolerability profile, and its efficacy in reducing HbA1c in monotherapy (120 mg before meals) is comparable to that of metformin, sulphonylureas, thiazolidinediones or acarbose (-0.5 to -1.5%). When combined with metformin, which primarily reduces fasting glucose levels, nateglinide's effects are additive. In our clinical experience, nateglinide is a particularly good therapeutic option in newly diagnosed, treatment-naive patients; elderly patients in whom hypoglycaemia is a concern; patients with kidney failure or mild hepatic impairment; patients taking low-dose sulphonylureas who encounter problems with hypoglycaemia; and patients failing to achieve adequate glycaemic control on metformin or thiazolidinedione monotherapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Nateglinide, ≥98% (HPLC), solid
USP
Nateglinide, United States Pharmacopeia (USP) Reference Standard
Nateglinide, European Pharmacopoeia (EP) Reference Standard