Merck
  • Systematic review and meta-analysis: D-Penicillamine vs. placebo/no intervention in patients with primary biliary cirrhosis--Cochrane Hepato-Biliary Group.
Systematic review and meta-analysis: D-Penicillamine vs. placebo/no intervention in patients with primary biliary cirrhosis--Cochrane Hepato-Biliary Group.
Alimentary pharmacology & therapeutics (2007-01-09)
Y Gong, S L Klingenberg, C Gluud
RESUMEN
D-Penicillamine is used for patients with primary biliary cirrhosis due to its ability to decrease hepatic copper and modulate the immune response. The results on effects of D--penicillamine in randomized-clinical trials of primary biliary cirrhosis patients are inconsistent. To systematically evaluate the benefits and harms of D-penicillamine for patients with primary biliary cirrhosis. We have performed a systematic review with meta-analyses of randomized-clinical trials to evaluate the effects of D-penicillamine for primary biliary cirrhosis. The primary outcomes are mortality and mortality or liver transplantation. We analysed the data by fixed-effect and random-effect models. Seven randomized trials including 706 patients were analysed. d-Penicillamine was without significant effects on mortality (RR 1.08, 95% CI: 0.82-1.43, P = 0.56), mortality or liver transplantation (RR 1.11, 95% CI: 0.74-1.68, P = 0.62), pruritus, liver complications, progression of liver histological stage and liver biochemical variables. D--Penicillamine significantly decreased serum alanine aminotransferase activity (weighted mean difference -45 IU/L, 95% CI: -75 to -15, P < 0.05) and led to significantly more adverse events (RR 4.18, 95% CI: 1.38-12.69, P = 0.01). D-Penicillamine did not appear to reduce the risk of mortality or morbidity, and led to more adverse events in patients with primary biliary cirrhosis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
D-Penicillamine, 98-101%
Penicillamine, European Pharmacopoeia (EP) Reference Standard