- Amiodarone supplants lidocaine in ACLS and CPR protocols.
Amiodarone supplants lidocaine in ACLS and CPR protocols.
Anesthesiology clinics (2011-08-30)
Anna Mizzi, Thanh Tran, Devanand Mangar, Enrico M Camporesi
PMID21871409
RESUMEN
Amiodarone is an antiarrhythmic medication used to treat and prevent certain types of serious, life-threatening ventricular arrhythmias. Amiodarone gained slow acceptance outside the specialized field of cardiac antiarrhythmic surgery because the side-effects are significant. Recent adoption of amiodarone in the ACLS (Advanced Cardiac Life Support) protocol has somewhat popularized this class of antiarrhythmics. Its use is slowly expanding in the acute medicine setting of anesthetics. This article summarizes the use of Amiodarone by anesthesiologists in the operating room and during cardiopulmonary resuscitation.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Lidocaine, European Pharmacopoeia (EP) Reference Standard