Merck
Erythropoietin.
Cold Spring Harbor perspectives in medicine (2013-03-05)
H Franklin Bunn
RESUMEN
During the past century, few proteins have matched erythropoietin (Epo) in capturing the imagination of physiologists, molecular biologists, and, more recently, physicians and patients. Its appeal rests on its commanding role as the premier erythroid cytokine, the elegant mechanism underlying the regulation of its gene, and its remarkable impact as a therapeutic agent, arguably the most successful drug spawned by the revolution in recombinant DNA technology. This concise review will begin with a synopsis of the colorful history of this protein, culminating in its purification and molecular cloning. It then covers in more detail the contemporary understanding of Epo's physiology as well as its structure and interaction with its receptor. A major part of this article focuses on the regulation of the Epo gene and the discovery of HIF, a transcription factor that plays a cardinal role in molecular adaptation to hypoxia. In the concluding section, a synopsis of Epo's role in disorders of red blood cell production will be followed by an assessment of the remarkable impact of Epo therapy in the treatment of anemias, as well as concerns that provide a strong impetus for the development of even safer and more effective treatment.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Erythropoietin human, EPO, recombinant, expressed in HEK 293 cells, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Erythropoietin, BRP, European Pharmacopoeia (EP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad