Merck
Adverse consequences of lysergic acid diethylamide.
Addiction (Abingdon, England) (1993-10-01)
H D Abraham, A M Aldridge
RESUMEN
The continued endemic use of hallucinogenic drugs, and of LSD in particular, raises concern regarding their short and long term adverse consequences. The epidemiology of LSD abuse is reviewed suggesting an increase in LSD use among the young as the prevalence rates for other substances continues to fall. Evidence supports the association of LSD use with panic reactions, prolonged schizoaffective psychoses and post-hallucinogen perceptual disorder, the latter being present continually for as long as 5 years. Evidence does not support claims of genetic disorders arising from hallucinogens. In light of the foregoing, current data confirm earlier findings of long lasting psychopathology arising in vulnerable individuals from the use of LSD. A hypothetical long term molecular mechanism of adverse effects is proposed.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
LSD solution, 25 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
LSD solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Lysergic acid diethylamide
En este momento no podemos mostrarle ni los precios ni la disponibilidad