Merck
  • Rituximab-cyclophosphamide-dexamethasone is highly effective in patients with monoclonal Ig deposit-related glomerulopathy and indolent non-Hodgkin lymphomas.
Rituximab-cyclophosphamide-dexamethasone is highly effective in patients with monoclonal Ig deposit-related glomerulopathy and indolent non-Hodgkin lymphomas.
American journal of hematology (2014-07-22)
Marielle Perry, Antoine Delarche, David Ribes, François Vergez, Céline Guilbeau-Frugier, Camille Laurent, Antoine Huart, Suzanne Tavitian, Hélène El Hachem, Lucie Oberic, Dominique Chauveau, Anne-Sophie Michallet, Loïc Ysebaert, Stanislas Faguer
RESUMEN
Indolent non-hodgkin lymphomas (iNHL) are a rare cause of monoclonal immunoglobulin deposits-related glomerulopathy (mIgGN). In patients with iNHL-related mIgGN, whether treatment should include either single or a combination of drug(s) to target the malignant clone and renal inflammation remains elusive. In this retrospective study, we report a cohort of 14 patients with iNHL-related mIgGN (cryoglobulinemic glomerulonephritis [n = 5], membranous nephropathy [n = 3], membranoproliferative glomerulonephritis [n = 3], AL or AL/AH amyloidosis [n = 2], and Light Chain Deposits Disease [n = 1]) and who received a treatment combining rituximab, cyclophosphamide, and dexamethasone (RCD). After a mean follow-up of 18 ± 4 months, nine patients (63%) had complete haematological response. Renal response was observed in 12 of the 14 patients (86%; complete response: n = 9; partial: n = 3). Estimated glomerular filtration rate increased from 47 ± 7 to 63 ± 8 mL/min/1.73 m(2) , and proteinuria decreased from 6.5 ± 0.7 to 1.4 ± 0.8 g/24 hr at one year. Following hematological relapse, renal relapse occurred in two patients suggesting sustained clonal eradication offers the best renal protection. Tolerance of RCD was good and the most frequent adverse event was pneumonia (3/14, 21%). RCD is a promising regimen for patients with iNHL and mIgGN, irrespective of glomerular pathologic pattern. Whether steroids can be avoided or minimized remains to be addressed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dexametasona, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexametasona, ≥98% (HPLC), powder
Supelco
Dexametasona, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexametasona, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dexametasona, meets USP testing specifications
Dexametasona, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexametasona, VETRANAL®, analytical standard
Sigma-Aldrich
Dexametasona, tested according to Ph. Eur.
Dexametasona, European Pharmacopoeia (EP) Reference Standard
Dexametasona, European Pharmacopoeia (EP) Reference Standard