Merck
Assessment of multiple hormonal activities in wastewater at different stages of treatment.
Environmental toxicology and chemistry (2014-07-01)
Peter A Bain, Mike Williams, Anu Kumar
RESUMEN
Changes in the endocrine potency of municipal wastewater at 3 wastewater treatment plants (WWTPs) in Australia were investigated using a panel of in vitro receptor-driven transactivation assays. The assays were based on human estrogen receptor α, androgen receptor, progesterone receptor, glucocorticoid receptor, and peroxisome proliferator-activated receptor γ2. Total removal efficiencies for estrogenic activity in the dissolved phase were 79.8% to 99.4%. Chemical analysis of 17β-estradiol, estrone, and 17α-ethinylestradiol levels showed that they accounted for the majority of the observed in vitro estrogenic activity in the final effluents but only 18% to 70% of estrogenic activity in the influents. Removal efficiency for androgenic activity was 97.5% to 100%. Endocrine activity levels were low in the final effluent of the WWTP with the lowest catchment population, with only estrogenic activity detected. In the final effluent of the WWTP with an intermediate catchment population, estrogenic, glucocorticoid, and peroxisome proliferator activities were detected. Estrogenic, antiandrogenic, progestagenic, glucocorticoid, and peroxisome proliferator activities were detected in the final effluent of the WWTP with the highest catchment population. The present study confirms the efficacy of secondary and tertiary treatment in reducing the concentrations of endocrine-active compounds in municipal wastewater. Further work is required to determine the possible health risks to aquatic biota posed by multiple hormonal activities present at low levels.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
17α-etinilestradiol, ≥98%
Sigma-Aldrich
Estrone, ≥99%
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
17α-etinilestradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
β-Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Estradiol, analytical standard
USP
β-Estradiol, United States Pharmacopeia (USP) Reference Standard
USP
Etinil estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Estrone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
17α-etinilestradiol, European Pharmacopoeia (EP) Reference Standard
β-Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
17α-etinilestradiol, European Pharmacopoeia (EP) Reference Standard
Estrone, European Pharmacopoeia (EP) Reference Standard