Merck
Antimicrobial activity of fullerenes and their hydroxylated derivatives.
Biocontrol science (2009-07-08)
Hisae Aoshima, Ken Kokubo, Shogo Shirakawa, Masayuki Ito, Shuichi Yamana, Takumi Oshima
RESUMEN
The antimicrobial activities of fullerene C60 and its derivatives against 6 kinds of bacteria and 2 kinds of fungi were evaluated. The tested samples were water-soluble fullerenes (polyvinylpyrrolidone (PVP)/C60, gamma-cyclodextrin (gamma-CD)/C60, and nano-C60) and 3 types of fullerenols (C60(OH)12, C60(OH)36.8H2O, and C60(OH)44.8H2O). Their activities were compared with those of (+)-catechin and hinokitiol from the viewpoint of future application to cosmetics. Although pristine C60 demonstrated no antimicrobial activity, fullerenols exhibited good antimicrobial activity against Propionibacterium acnes, Staphylococcus epidermidis, Candida albicans, and Malassezia furfur. In particular, C60(OH)44 exhibited a strong and wide-ranging antimicrobial activity comparable to that of catechin. This compound exhibits antimicrobial activity via inhibition of microbial cell growth and not via bactericidal activity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Polyhydroxylated fullerenes, water soluble